COM448612768 Combi-Blocks, Inc.

1-(5-Aminoquinolin-6-yl)piperidin-4-ol

  • MDL number MFCD12067790
  • Cas Number : 1157642-42-6
  • Molecular Weight : 243.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3801

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3801
Availability details for
Quantity
  • Availability
  • Add to Cart