COM448612770 Combi-Blocks, Inc.

3-(Piperidin-4-yloxy)aniline, HCl

  • MDL number MFCD16620352
  • Cas Number : 1370365-89-1
  • Molecular Weight : 228.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3803

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3803
Availability details for
Quantity
  • Availability
  • Add to Cart