COM448612771 Combi-Blocks, Inc.

1-N-Benzyl-2-bromobenzene-1,4-diamine

  • MDL number MFCD16085336
  • Cas Number : 1248984-11-3
  • Molecular Weight : 277.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3804

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3804
Availability details for
Quantity
  • Availability
  • Add to Cart