COM448612773 Combi-Blocks, Inc.

2-Bromo-1-N-methylbenzene-1,4-diamine

  • MDL number MFCD16085328
  • Cas Number : 1248092-75-2
  • Molecular Weight : 201.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3806

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3806
Availability details for
Quantity
  • Availability
  • Add to Cart