COM448612774 Combi-Blocks, Inc.

2-Bromo-1-N-ethylbenzene-1,4-diamine

  • MDL number MFCD16085330
  • Cas Number : 1247648-17-4
  • Molecular Weight : 215.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3807

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3807
Availability details for
Quantity
  • Availability
  • Add to Cart