COM448612775 Combi-Blocks, Inc.

2-Bromo-1-N,1-N-diethylbenzene-1,4-diamine

  • MDL number MFCD16085281
  • Cas Number : 105294-26-6
  • Molecular Weight : 243.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3808

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3808
Availability details for
Quantity
  • Availability
  • Add to Cart