COM448612776 Combi-Blocks, Inc.

2-Bromo-1-N-propylbenzene-1,4-diamine

  • MDL number MFCD16085318
  • Cas Number : 1250988-00-1
  • Molecular Weight : 229.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3809

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3809
Availability details for
Quantity
  • Availability
  • Add to Cart