COM448612784 Combi-Blocks, Inc.

4-Chloro-2-iodo-5-methylaniline

  • MDL number MFCD22192403
  • Cas Number : 1373233-50-1
  • Molecular Weight : 267.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3818

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3818
Availability details for
Quantity
  • Availability
  • Add to Cart