COM448612788 Combi-Blocks, Inc.

Tert-butyl 4-(2-amino-5-methoxyphenyl)piperazine-1-carboxylate

  • MDL number MFCD22192406
  • Cas Number : 1373232-41-7
  • Molecular Weight : 307.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3822

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3822
Availability details for
Quantity
  • Availability
  • Add to Cart