COM448612791 Combi-Blocks, Inc.

2-(4-Amino-2,6-dibromophenoxy)acetonitrile

  • MDL number MFCD11647736
  • Cas Number : 1094566-60-5
  • Molecular Weight : 306.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3825

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3825
Availability details for
Quantity
  • Availability
  • Add to Cart