COM448612792 Combi-Blocks, Inc.

2-Amino-4-trifluoromethylphenyl disulfide

  • MDL number MFCD22192407
  • Cas Number : 7038-40-6
  • Molecular Weight : 384.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3826

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3826
Availability details for
Quantity
  • Availability
  • Add to Cart