COM448612796 Combi-Blocks, Inc.

(Z)-[Amino(4-aminophenyl)methylidene]amino 4-chlorobenzoate

  • MDL number MFCD22192410
  • Cas Number : 1373237-01-4
  • Molecular Weight : 289.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3830

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3830
Availability details for
Quantity
  • Availability
  • Add to Cart