COM448612801 Combi-Blocks, Inc.

4-(5-Ethyl-1,2,4-oxadiazol-3-yl)aniline

  • MDL number MFCD12191340
  • Cas Number : 10185-71-4
  • Molecular Weight : 189.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3835

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3835
Availability details for
Quantity
  • Availability
  • Add to Cart