COM448612805 Combi-Blocks, Inc.

4-(5-Butyl-1,2,4-oxadiazol-3-yl)aniline

  • MDL number MFCD12191346
  • Cas Number : 10364-73-5
  • Molecular Weight : 217.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3839

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3839
Availability details for
Quantity
  • Availability
  • Add to Cart