COM448612806 Combi-Blocks, Inc.

4-[5-(1-Chloro-2-methylpropan-2-yl)-1,2,4-oxadiazol-3-yl]aniline

  • MDL number MFCD22192411
  • Cas Number : 1373233-51-2
  • Molecular Weight : 251.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3840

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3840
Availability details for
Quantity
  • Availability
  • Add to Cart