COM448612807 Combi-Blocks, Inc.

4-(5-Cyclohexyl-1,2,4-oxadiazol-3-yl)aniline

  • MDL number MFCD12426365
  • Cas Number : 96898-79-2
  • Molecular Weight : 243.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3841

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3841
Availability details for
Quantity
  • Availability
  • Add to Cart