COM448612809 Combi-Blocks, Inc.

3-(5-Cyclohexyl-1,2,4-oxadiazol-3-yl)aniline

  • MDL number MFCD09259053
  • Cas Number : 929338-71-6
  • Molecular Weight : 243.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3843

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3843
Availability details for
Quantity
  • Availability
  • Add to Cart