COM448612810 Combi-Blocks, Inc.

3-[5-(Chloromethyl)-1,2,4-oxadiazol-3-yl]aniline

  • MDL number MFCD07369927
  • Cas Number : 6595-79-5
  • Molecular Weight : 209.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3844

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3844
Availability details for
Quantity
  • Availability
  • Add to Cart