COM448612814 Combi-Blocks, Inc.

3-[3-(4-Bromophenyl)-1,2,4-oxadiazol-5-yl]aniline

  • MDL number MFCD09034356
  • Cas Number : 937654-87-0
  • Molecular Weight : 316.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3848

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3848
Availability details for
Quantity
  • Availability
  • Add to Cart