COM448612817 Combi-Blocks, Inc.

3-[5-(2-Methoxyphenyl)-1,2,4-oxadiazol-3-yl]aniline

  • MDL number MFCD09259050
  • Cas Number : 929338-65-8
  • Molecular Weight : 267.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3851

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3851
Availability details for
Quantity
  • Availability
  • Add to Cart