COM448612824 Combi-Blocks, Inc.

1-(5-Amino-2-methylimidazol-1-yl)propan-2-ol

  • MDL number MFCD20720185
  • Cas Number : 242132-98-5
  • Molecular Weight : 155.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3865

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3865
Availability details for
Quantity
  • Availability
  • Add to Cart