COM448612835 Combi-Blocks, Inc.

4-Amino-2,3-dimethylpyridin-1-ium-1-olate

  • MDL number MFCD00065173
  • Cas Number : 156118-22-8
  • Molecular Weight : 138.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3884

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3884
Availability details for
Quantity
  • Availability
  • Add to Cart