COM448612837 Combi-Blocks, Inc.

2,5-Diaminobenzoic acid

  • MDL number MFCD00127689
  • Cas Number : 610-74-2
  • Molecular Weight : 152.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3887

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3887
Availability details for
Quantity
  • Availability
  • Add to Cart