COM448612838 Combi-Blocks, Inc.

5-(4-Aminophenyl)furan-2-carbaldehyde

  • MDL number MFCD20697214
  • Cas Number : 57023-90-2
  • Molecular Weight : 187.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3889

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3889
Availability details for
Quantity
  • Availability
  • Add to Cart