COM448612843 Combi-Blocks, Inc.

3-Amino-4-[(2-hydroxyethyl)amino]phenol

  • MDL number MFCD22192416
  • Cas Number : 1373232-77-9
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3895

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3895
Availability details for
Quantity
  • Availability
  • Add to Cart