COM448612854 Combi-Blocks, Inc.

4-Amino-2-methylpyridin-1-ium-1-olate

  • MDL number MFCD01646157
  • Cas Number : 14045-17-1
  • Molecular Weight : 124.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3910

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3910
Availability details for
Quantity
  • Availability
  • Add to Cart