COM448612866 Combi-Blocks, Inc.

3-Amino-4-bromo-N-methylbenzamide

  • MDL number MFCD11938549
  • Cas Number : 1177098-90-6
  • Molecular Weight : 229.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3925

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3925
Availability details for
Quantity
  • Availability
  • Add to Cart