COM448612870 Combi-Blocks, Inc.

3-Amino-4-(tert-butylamino)benzonitrile

  • MDL number MFCD12189583
  • Cas Number : 320406-79-9
  • Molecular Weight : 189.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3929

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3929
Availability details for
Quantity
  • Availability
  • Add to Cart