COM448612873 Combi-Blocks, Inc.

1-N-Isopropyl-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1,2-diamine

  • MDL number MFCD22192422
  • Cas Number : 2096998-67-1
  • Molecular Weight : 276.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3933

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3933
Availability details for
Quantity
  • Availability
  • Add to Cart