COM448612882 Combi-Blocks, Inc.

Methyl 2-[(2Z)-5-amino-1H-pyridin-2-ylidene]-2-cyanoacetate

  • MDL number MFCD22192427
  • Cas Number : 1373237-02-5
  • Molecular Weight : 191.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3942

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3942
Availability details for
Quantity
  • Availability
  • Add to Cart