COM448612883 Combi-Blocks, Inc.

5-Amino-3-bromopyridine-2-thiol

  • MDL number MFCD21946602
  • Cas Number : 1373232-57-5
  • Molecular Weight : 205.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3943

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3943
Availability details for
Quantity
  • Availability
  • Add to Cart