COM448612886 Combi-Blocks, Inc.

1,3-Dimethyl 2-(3-amino-5-bromopyridin-2-yl)propanedioate

  • MDL number MFCD22192428
  • Cas Number : 1373233-26-1
  • Molecular Weight : 303.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3946

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3946
Availability details for
Quantity
  • Availability
  • Add to Cart