COM448612887 Combi-Blocks, Inc.

Tert-butyl 2-[(2E)-5-amino-1H-pyridin-2-ylidene]-2-cyanoacetate

  • MDL number MFCD22192429
  • Cas Number : 1373237-03-6
  • Molecular Weight : 233.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3947

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3947
Availability details for
Quantity
  • Availability
  • Add to Cart