COM448612892 Combi-Blocks, Inc.

5-Amino-3-methylpyridin-2-ol

  • MDL number MFCD07375119
  • Cas Number : 20252-07-7
  • Molecular Weight : 124.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3952

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3952
Availability details for
Quantity
  • Availability
  • Add to Cart