COM448612893 Combi-Blocks, Inc.

Tert-butyl 2-(3-aminopyridin-2-yl)-2-cyanoacetate

  • MDL number MFCD22192431
  • Cas Number : 1373233-08-9
  • Molecular Weight : 233.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3953

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3953
Availability details for
Quantity
  • Availability
  • Add to Cart