COM448612895 Combi-Blocks, Inc.

Bis(5-Aminopyridin-2-yl)sulfane

  • MDL number MFCD00038834
  • Cas Number : 32605-02-0
  • Molecular Weight : 218.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3955

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3955
Availability details for
Quantity
  • Availability
  • Add to Cart