COM448612897 Combi-Blocks, Inc.

6-Chloro-2-N-methylpyridine-2,3-diamine

  • MDL number MFCD11047119
  • Cas Number : 89660-14-0
  • Molecular Weight : 157.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3957

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3957
Availability details for
Quantity
  • Availability
  • Add to Cart