COM448612903 Combi-Blocks, Inc.

6-Bromo-[1,2,4]triazolo[4,3-a]pyridin-8-amine

  • MDL number MFCD14545816
  • Cas Number : 1216292-05-5
  • Molecular Weight : 213.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3963

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3963
Availability details for
Quantity
  • Availability
  • Add to Cart