COM448612906 Combi-Blocks, Inc.

5-Bromo-2-(pyrrolidin-1-yl)pyridin-3-amine

  • MDL number MFCD14600434
  • Cas Number : 1226063-82-6
  • Molecular Weight : 242.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3966

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3966
Availability details for
Quantity
  • Availability
  • Add to Cart