COM448612910 Combi-Blocks, Inc.

5-Bromo-2-N-cyclopropylpyridine-2,3-diamine

  • MDL number MFCD08692116
  • Cas Number : 1010422-25-9
  • Molecular Weight : 228.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3970

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3970
Availability details for
Quantity
  • Availability
  • Add to Cart