COM448612911 Combi-Blocks, Inc.

2-[(3-Amino-5-bromopyridin-2-yl)amino]ethanol

  • MDL number MFCD14543349
  • Cas Number : 1216024-64-4
  • Molecular Weight : 232.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3971

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3971
Availability details for
Quantity
  • Availability
  • Add to Cart