COM448612914 Combi-Blocks, Inc.

5-Bromo-2-(piperidin-1-yl)pyridin-3-amine

  • MDL number MFCD14600443
  • Cas Number : 1226271-22-2
  • Molecular Weight : 256.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3974

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3974
Availability details for
Quantity
  • Availability
  • Add to Cart