COM448612921 Combi-Blocks, Inc.

2-N-Isopropyl-5-methylpyridine-2,3-diamine

  • MDL number MFCD14543395
  • Cas Number : 1216221-51-0
  • Molecular Weight : 165.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3981

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3981
Availability details for
Quantity
  • Availability
  • Add to Cart