COM448612922 Combi-Blocks, Inc.

2-N-Ethyl-5-methylpyridine-2,3-diamine

  • MDL number MFCD14543397
  • Cas Number : 1215946-13-6
  • Molecular Weight : 151.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3982

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3982
Availability details for
Quantity
  • Availability
  • Add to Cart