COM448612924 Combi-Blocks, Inc.

4-Amino-2-chloro-5-methylpyridin-1-ium-1-olate

  • MDL number MFCD22192434
  • Cas Number : 1373232-42-8
  • Molecular Weight : 158.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3984

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3984
Availability details for
Quantity
  • Availability
  • Add to Cart