COM448612935 Combi-Blocks, Inc.

Potassium [3-amino-5-(methoxycarbonyl)phenyl]trifluoroboranuide

  • MDL number MFCD22192435
  • Molecular Weight : 257.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3996

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3996
Availability details for
Quantity
  • Availability
  • Add to Cart