COM448612939 Combi-Blocks, Inc.

4-(4-Nitrophenyl)thiomorpholine

  • MDL number MFCD01604442
  • Cas Number : 90254-22-1
  • Molecular Weight : 224.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-4000

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-4000
Availability details for
Quantity
  • Availability
  • Add to Cart