COM448612950 Combi-Blocks, Inc.

2-Fluoro-3-(trifluoroacetamido)phenylboronic acid

  • MDL number MFCD28369556
  • Cas Number : 2377610-93-8
  • Molecular Weight : 250.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6019

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6019
Availability details for
Quantity
  • Availability
  • Add to Cart