COM448612957 Combi-Blocks, Inc.

N-(5-Bromo-2-nitrophenyl)-2,2,2-trifluoroacetamide

  • MDL number MFCD28385190
  • Cas Number : 1820711-51-0
  • Molecular Weight : 313.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6028

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6028
Availability details for
Quantity
  • Availability
  • Add to Cart