COM448612973 Combi-Blocks, Inc.

(Oxan-4-ylmethyl)boronic acid

  • MDL number MFCD18861659
  • Cas Number : 1350513-18-6
  • Molecular Weight : 144

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0005

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-0005
Availability details for
Quantity
  • Availability
  • Add to Cart